Alfa Aesar™ 2-Chloro-6-fluorophenol, 97%

Manufacturer: Alfa Aesar™ B2330303

Quantity 1g

 View more versions of this product

Catalog No. AAB2330303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2040-90-6
    Molecular Formula C6H4ClFO
    Formula Weight 146.55
    MDL Number MFCD01631574
    UN Number UN3261
    Beilstein 1936441
    Chemical Name or Material 2-Chloro-6-fluorophenol

    Specifications

    Formula Weight 146.55
    UN Number UN3261
    Melting Point 61° to 65°C
    Quantity 1g
    Molecular Formula C6H4ClFO
    MDL Number MFCD01631574
    CAS 2040-90-6
    Beilstein 1936441
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H340-H361-H314-H318-H302-H317
    May cause genetic defects.
    Suspected of damaging fertility or the unborn child.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P273-P201-P309-P310
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Odor Phenol-like
    Chemical Name or Material 2-Chloro-6-fluorophenol
    Assay 97%

    GHS H Statement
    H340-H361-H314-H318-H302-H317
    May cause genetic defects.
    Suspected of damaging fertility or the unborn child.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P273-P201-P309-P310
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit