Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-6-fluorophenol, 97%

Click to view available options
Quantity:
1g
5g
2040-90-6
C6H4ClFO
146.545
MFCD01631574
QIAQIYQASAWZPP-UHFFFAOYSA-N
2-chloro-6-fluoro-phenol,2-chloro-6-fluoro phenol,phenol, 2-chloro-6-fluoro,pubchem2853,6-chlor-2-fluorphenol,acmc-1cqou,6-chloro-2-fluorophenol,2-chlor-6-fluorphenol;,2-chloro-6-fluorophenol;,2-chlor-6-fluor-phenol;
2773710
2-chloro-6-fluorophenol
C1=CC(=C(C(=C1)Cl)O)F
2040-90-6
C6H4ClFO
146.545
MFCD01631574
QIAQIYQASAWZPP-UHFFFAOYSA-N
2-chloro-6-fluoro-phenol,2-chloro-6-fluoro phenol,phenol, 2-chloro-6-fluoro,pubchem2853,6-chlor-2-fluorphenol,acmc-1cqou,6-chloro-2-fluorophenol,2-chlor-6-fluorphenol;,2-chloro-6-fluorophenol;,2-chlor-6-fluor-phenol;
2773710
2-chloro-6-fluorophenol
C1=CC(=C(C(=C1)Cl)O)F
Quantity 5g
Odor Phenol-like
Melting Point 61°C to 65°C
UN Number UN3261
Beilstein 1936441
Formula Weight 146.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorophenol

GHS H Statement
H340-H361-H314-H318-H302-H317
May cause genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P273-P201-P309-P310
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.