Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-fluorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
313545-32-3
C6H5BClFO2
174.37
NXSZSZJWZVLHAY-UHFFFAOYSA-N
2-chloro-6-fluorophenyl boronic acid,2-chloro-6-fluorobenzeneboronic acid,2-chloro-6-fluorophenylboronicacid,boronic acid, 2-chloro-6-fluorophenyl,boronic acid,b-2-chloro-6-fluorophenyl,boronic acid, b-2-chloro-6-fluorophenyl,pubchem18883,acmc-1ad0v,2-borono-1-chloro-3-fluorobenzene,2-chloro-6-fluorobenzene boronic acid
3549395
(2-chloro-6-fluorophenyl)boronic acid
B(C1=C(C=CC=C1Cl)F)(O)O
313545-32-3
C6H5BClFO2
174.37
NXSZSZJWZVLHAY-UHFFFAOYSA-N
2-chloro-6-fluorophenyl boronic acid,2-chloro-6-fluorobenzeneboronic acid,2-chloro-6-fluorophenylboronicacid,boronic acid, 2-chloro-6-fluorophenyl,boronic acid,b-2-chloro-6-fluorophenyl,boronic acid, b-2-chloro-6-fluorophenyl,pubchem18883,acmc-1ad0v,2-borono-1-chloro-3-fluorobenzene,2-chloro-6-fluorobenzene boronic acid
3549395
(2-chloro-6-fluorophenyl)boronic acid
B(C1=C(C=CC=C1Cl)F)(O)O
Melting Point 129.9°C to 132.8°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 174.37
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-chloro-6-fluorophenylboronic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning