Alfa Aesar™ 2-Chloro-6-iodotoluene, 98%

Manufacturer: Alfa Aesar™ A1158206

Quantity 5g

 View more versions of this product

Catalog No. AAA1158206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42048-11-3
    Molecular Formula C7H6ClI
    Formula Weight 252.48
    MDL Number MFCD00019013
    Beilstein 2433704
    Synonym 1-Chloro-3-iodo-2-methylbenzene; 3-Chloro-2-methyliodobenzene
    Chemical Name or Material 2-Chloro-6-iodotoluene

    Specifications

    Formula Weight 252.48
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H6ClI
    Beilstein 2433704
    Boiling Point 119° to 120°C (15mmHg)
    Melting Point 11°C
    Quantity 5g
    MDL Number MFCD00019013
    Synonym 1-Chloro-3-iodo-2-methylbenzene; 3-Chloro-2-methyliodobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 42048-11-3
    Chemical Name or Material 2-Chloro-6-iodotoluene
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit