Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-6-methoxybenzothiazole, 97%

Manufacturer:  Alfa Aesar™ H3277006

Quantity 5g

 View more versions of this product

Catalog No. AAH3277006


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2605-14-3
199.66
2-Chloro-6-methoxybenzothiazole
C8H6ClNOS
MFCD00671510
2-Chloro-6-methoxybenzothiazole

Specifications

52° to 54°C
2605-14-3
199.66
2-Chloro-6-methoxybenzothiazole
97%
5g
C8H6ClNOS
MFCD00671510
2-Chloro-6-methoxybenzothiazole

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit