Alfa Aesar™ 2-Chloro-6-methoxynicotinic acid, 95%

Manufacturer: Alfa Aesar™ H3400103

Quantity 1g

 View more versions of this product

Catalog No. AAH3400103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 503000-87-1
    Molecular Formula C7H6ClNO3
    Formula Weight 187.6
    MDL Number MFCD08236845
    Synonym 2-Chloro-6-methoxypyridine-3-carboxylic acid
    Chemical Name or Material 2-Chloro-6-methoxynicotinic acid

    Specifications

    Formula Weight 187.6
    Molecular Formula C7H6ClNO3
    MDL Number MFCD08236845
    CAS 503000-87-1
    Synonym 2-Chloro-6-methoxypyridine-3-carboxylic acid
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 231° to 234°C
    Quantity 1g
    Health Hazard 1 Warning
    Chemical Name or Material 2-Chloro-6-methoxynicotinic acid
    Assay 95%

    Description

    Applications
    2-Chloro-6-methylpyridine-3-carboxylic acid (2-Chloro-6-methylnicotinic acid) may be used in the synthesis of 6-methyl-2-thiophenoxynicotinic acid, required for the preparation of europium complex for staining the nucleolus of NIH 3T3, HeLa and HDF cells.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit