Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-6-methyl-4-nitroaniline, 98%

Click to view available options
Quantity:
1g
5g
69951-02-6
C7H7ClN2O2
186.595
MFCD01313703
HVPVUAIKGOKEEE-UHFFFAOYSA-N
2-chloro-4-nitro-6-methylaniline,2-chloro-6-methyl-4-nitro-phenylamine,benzenamine,2-chloro-6-methyl-4-nitro,benzenamine, 2-chloro-6-methyl-4-nitro,6-chloro-2-methyl-4-nitrophenylamine,pubchem4373,chloromethylnitroaniline,acmc-1b462,6-chloro-2-methyl-4-nitroaniline,2-chloro-6-methyl-4-nitrophenylamine
2733460
2-chloro-6-methyl-4-nitroaniline
CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-]
69951-02-6
C7H7ClN2O2
186.595
MFCD01313703
HVPVUAIKGOKEEE-UHFFFAOYSA-N
2-chloro-4-nitro-6-methylaniline,2-chloro-6-methyl-4-nitro-phenylamine,benzenamine,2-chloro-6-methyl-4-nitro,benzenamine, 2-chloro-6-methyl-4-nitro,6-chloro-2-methyl-4-nitrophenylamine,pubchem4373,chloromethylnitroaniline,acmc-1b462,6-chloro-2-methyl-4-nitroaniline,2-chloro-6-methyl-4-nitrophenylamine
2733460
2-chloro-6-methyl-4-nitroaniline
CC1=CC(=CC(=C1N)Cl)[N+](=O)[O-]
Melting Point 174°C to 176°C
Quantity 1g
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methyl-4-nitroaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger