Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-methylpyrimidine-4-carboxylic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
89581-58-8
C6H5ClN2O2
172.57
MFCD00833002
FBXGLOGQWMGIER-UHFFFAOYSA-N
2-chloro-6-methyl-pyrimidine-4-carboxylic acid,2-chloro-6-methyl-4-pyrimidinecarboxylic acid,2-chloro-4-methylpyrimidine-6-carboxylic acid,4-pyrimidinecarboxylic acid, 2-chloro-6-methyl
10535213
2-chloro-6-methylpyrimidine-4-carboxylic acid
CC1=NC(=NC(=C1)C(=O)O)Cl
89581-58-8
C6H5ClN2O2
172.57
MFCD00833002
FBXGLOGQWMGIER-UHFFFAOYSA-N
2-chloro-6-methyl-pyrimidine-4-carboxylic acid,2-chloro-6-methyl-4-pyrimidinecarboxylic acid,2-chloro-4-methylpyrimidine-6-carboxylic acid,4-pyrimidinecarboxylic acid, 2-chloro-6-methyl
10535213
2-chloro-6-methylpyrimidine-4-carboxylic acid
CC1=NC(=NC(=C1)C(=O)O)Cl
Melting Point 172°C to 173°C
Quantity 5g
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 172.57
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methylpyrimidine-4-carboxylic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning