Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-nitrobenzaldehyde, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
6361-22-4
C7H4ClNO3
185.57
RZDOUWDCYULHJX-UHFFFAOYSA-N
2-nitro-6-chlorobenzaldehyde,benzaldehyde, 2-chloro-6-nitro,6-chloro-2-nitrobenzaldehyde,pubchem17009,2-chloro-6-nitro-benzaldehye,ksc497q1l,2-chloro-6-nitro-benzaldehyde,benzaldehyde,2-chloro-6-nitro,rzdouwdcyulhjx-uhfffaoysa,attercop-chm at114429
80701
2-chloro-6-nitrobenzaldehyde
C1=CC(=C(C(=C1)Cl)C=O)[N+](=O)[O-]
6361-22-4
C7H4ClNO3
185.57
RZDOUWDCYULHJX-UHFFFAOYSA-N
2-nitro-6-chlorobenzaldehyde,benzaldehyde, 2-chloro-6-nitro,6-chloro-2-nitrobenzaldehyde,pubchem17009,2-chloro-6-nitro-benzaldehye,ksc497q1l,2-chloro-6-nitro-benzaldehyde,benzaldehyde,2-chloro-6-nitro,rzdouwdcyulhjx-uhfffaoysa,attercop-chm at114429
80701
2-chloro-6-nitrobenzaldehyde
C1=CC(=C(C(=C1)Cl)C=O)[N+](=O)[O-]
Quantity 1g
Packaging Glass bottle
Melting Point 69.0°C to 72.0°C
Assay Percent Range 98%
Linear Formula ClC6H3(NO2)CHO
Beilstein 07,262
Formula Weight 185.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-nitrobenzaldehyde