Alfa Aesar™ 2-Chloro-6-(trifluoromethyl)benzoyl chloride, 97%

Manufacturer: Alfa Aesar™ H3006206

Quantity 5g

 View more versions of this product

Catalog No. AAH3006206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 916420-44-5
    Molecular Formula C8H3Cl2F3O
    Formula Weight 243.01
    MDL Number MFCD09025361
    UN Number UN3265
    Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzoyl chloride

    Specifications

    Formula Weight 243.01
    MDL Number MFCD09025361
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 5g
    Molecular Formula C8H3Cl2F3O
    CAS 916420-44-5
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzoyl chloride
    Assay 97%

    Description

    Applications
    The use of pentafluoropropionic anhydride, trifluoromethylbenzoyl chloride and pentafluorobenzoyl chloride gave fluoro-chitin derivatives with 40, 10 and 5% substitution, respectively. Fluorination of chitin was achieved by facile homogenous reaction of chitin solution with diethyl amino sulfur trifluoride.

    Solubility
    Insoluble in water.

    Notes
    Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducind agents, bases, water, moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit