Alfa Aesar™ (+)-2-Chloro-L-phenylglycine, 95%

Manufacturer: Alfa Aesar™ H2877606

Quantity 5g

 View more versions of this product

Catalog No. AAH2877606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141315-50-6
    Molecular Formula C8H8ClNO2
    Formula Weight 185.61
    MDL Number MFCD03426346
    Synonym H-Phg(2-Cl)-OH; H-Phg(2-Cl)-OH
    Chemical Name or Material (+)-2-Chloro-L-phenylglycine

    Specifications

    Formula Weight 185.61
    Molecular Formula C8H8ClNO2
    MDL Number MFCD03426346
    CAS 141315-50-6
    Synonym H-Phg(2-Cl)-OH; H-Phg(2-Cl)-OH
    Melting Point 182° to 187°C
    Quantity 5g
    Optical Rotation Not completely soluble in Chloroform
    Chemical Name or Material (+)-2-Chloro-L-phenylglycine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit