Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-N-(4-iodophenyl)acetamide, 95%, Thermo Scientific™

Catalog Number AC438290050
Click to view available options
Quantity:
5 g
2564-00-3
C8H7ClINO
295.51
WIUVQCIRRAAPAX-UHFFFAOYSA-N
2-chloro-n-4-iodophenyl acetamide, n1-4-iodophenyl-2-chloroacetamide, acetamide,2-chloro-n-4-iodophenyl, 4'-iodo-2-chloroacetoanilide, 2-chloro-n-4-iodo-phenyl-acetamide, acetamide, 2-chloro-n-4-iodophenyl, 10.14272/wiuvqcirraapax-uhfffaoysa-n, doi:10.14272/wiuvqcirraapax-uhfffaoysa-n
735792
2-chloro-N-(4-iodophenyl)acetamide
C1=CC(=CC=C1NC(=O)CCl)I
2564-00-3
C8H7ClINO
295.51
WIUVQCIRRAAPAX-UHFFFAOYSA-N
2-chloro-n-4-iodophenyl acetamide, n1-4-iodophenyl-2-chloroacetamide, acetamide,2-chloro-n-4-iodophenyl, 4'-iodo-2-chloroacetoanilide, 2-chloro-n-4-iodo-phenyl-acetamide, acetamide, 2-chloro-n-4-iodophenyl, 10.14272/wiuvqcirraapax-uhfffaoysa-n, doi:10.14272/wiuvqcirraapax-uhfffaoysa-n
735792
2-chloro-N-(4-iodophenyl)acetamide
C1=CC(=CC=C1NC(=O)CCl)I
CAS Min % 94.0
CAS Max % 100.0
Melting Point 194.8°C to 196.7°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 295.51
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-N-(4-iodophenyl)acetamide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit