Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-N-Fmoc-L-phenylalanine, 97%

Click to view available options
Quantity:
5g
25g
198560-41-7
C24H20ClNO4
421.877
MFCD00797578
RNNKPNPLIMFSDY-QFIPXVFZSA-N
fmoc-2-chloro-l-phenylalanine,fmoc-phe 2-cl-oh,fmoc-l-2-chlorophe,fmoc-l-2-chlorophenylalanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-chlorophenyl propanoic acid,2s-3-2-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,l-phenylalanine, 2-chloro-n-9h-fluoren-9-ylmethoxy carbonyl,2s-3-2-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-l-phe 2-cl-oh,ksc925c0b
2761464
(2S)-3-(2-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Cl
198560-41-7
C24H20ClNO4
421.877
MFCD00797578
RNNKPNPLIMFSDY-QFIPXVFZSA-N
fmoc-2-chloro-l-phenylalanine,fmoc-phe 2-cl-oh,fmoc-l-2-chlorophe,fmoc-l-2-chlorophenylalanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-chlorophenyl propanoic acid,2s-3-2-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,l-phenylalanine, 2-chloro-n-9h-fluoren-9-ylmethoxy carbonyl,2s-3-2-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-l-phe 2-cl-oh,ksc925c0b
2761464
(2S)-3-(2-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Cl
Beilstein 10203122
Formula Weight 421.88
Physical Form Powder
Percent Purity 97%
Color White
Quantity 5g
Chemical Name or Material 2-Chloro-N-Fmoc-L-phenylalanine

storageNote1 : 2° to 8°C