Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-methoxy-N-methylacetamide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
67442-07-3
C4H8ClNO2
137.56
MFCD00134232
SCOJKGRNQDKFRP-UHFFFAOYSA-N
2-chloro-n-methoxy-n-methyl acetamide,acetamide,2-chloro-n-methoxy-n-methyl,n-methoxy-n-methylchloroacetamide,acetamide, 2-chloro-n-methoxy-n-methyl,pubchem18657,acmc-209nz0,n-methyl-n-methoxy chloroacetamide,n-chloroacetyl-n-methoxymethylamine,n-methyl-n-methoxy-2-chloroacetamide,2-chloro-n-methoxy-n-methyl-acetamide
2734716
2-chloro-N-methoxy-N-methylacetamide
CN(C(=O)CCl)OC
67442-07-3
C4H8ClNO2
137.56
MFCD00134232
SCOJKGRNQDKFRP-UHFFFAOYSA-N
2-chloro-n-methoxy-n-methyl acetamide,acetamide,2-chloro-n-methoxy-n-methyl,n-methoxy-n-methylchloroacetamide,acetamide, 2-chloro-n-methoxy-n-methyl,pubchem18657,acmc-209nz0,n-methyl-n-methoxy chloroacetamide,n-chloroacetyl-n-methoxymethylamine,n-methyl-n-methoxy-2-chloroacetamide,2-chloro-n-methoxy-n-methyl-acetamide
2734716
2-chloro-N-methoxy-N-methylacetamide
CN(C(=O)CCl)OC
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 104°C
Melting Point 37.0°C to 42.0°C
Boiling Point 94.0°C to 95.0°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula ClCH2CON(OCH3)CH3
Formula Weight 137.56
Physical Form Crystals
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-methoxy-N-methylacetamide
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C