Alfa Aesar™ 2-Chloro-p-phenylenediamine sulfate, 95%

Manufacturer: Alfa Aesar™ B240290B

Quantity 1000g

 View more versions of this product

Catalog No. AAB240290B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61702-44-1
    Molecular Formula C6H7ClN2·H2SO4
    Formula Weight 240.67
    MDL Number MFCD00013004
    UN Number UN2811
    Beilstein 8441373
    Synonym 2-Chloro-1,4-diaminobenzene sulfate
    Chemical Name or Material 2-Chloro-p-phenylenediamine sulfate

    Specifications

    Formula Weight 240.67
    Molecular Formula C6H7ClN2·H2SO4
    MDL Number MFCD00013004
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 251° to 253°C
    Quantity 1000g
    CAS 61702-44-1
    Beilstein 8441373
    Synonym 2-Chloro-1,4-diaminobenzene sulfate
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number CZ1581000
    Chemical Name or Material 2-Chloro-p-phenylenediamine sulfate
    Assay 95%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit