Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-p-phenylenediamine sulfate, 95%

Click to view available options
Quantity:
250g
1000g
61702-44-1
C6H9ClN2O4S
240.658
MFCD00013004
GQFGHCRXPLROOF-UHFFFAOYSA-N
2-chloro-p-phenylenediamine sulfate,2-chlorobenzene-1,4-diamine sulfate,2-chloro-1,4-phenylenediamine sulfate,fourrine so,2-chloro-1,4-benzenediamine sulfate,2-chlorobenzene-1,4-diamine; sulfuric acid,1,4-benzenediamine, 2-chloro-, sulfate,renal so,fourrine 81,2-cl-p-pd
22584
CHEBI:76597
2-chlorobenzene-1,4-diamine;sulfuric acid
C1=CC(=C(C=C1N)Cl)N.OS(=O)(=O)O
61702-44-1
C6H9ClN2O4S
240.658
MFCD00013004
GQFGHCRXPLROOF-UHFFFAOYSA-N
2-chloro-p-phenylenediamine sulfate,2-chlorobenzene-1,4-diamine sulfate,2-chloro-1,4-phenylenediamine sulfate,fourrine so,2-chloro-1,4-benzenediamine sulfate,2-chlorobenzene-1,4-diamine; sulfuric acid,1,4-benzenediamine, 2-chloro-, sulfate,renal so,fourrine 81,2-cl-p-pd
22584
CHEBI:76597
2-chlorobenzene-1,4-diamine;sulfuric acid
C1=CC(=C(C=C1N)Cl)N.OS(=O)(=O)O
Melting Point 251°C to 253°C
Quantity 250g
UN Number UN2811
Beilstein 8441373
Formula Weight 240.67
Percent Purity 95%
Chemical Name or Material 2-Chloro-p-phenylenediamine sulfate

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CZ1581000