Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroanisole, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
766-51-8
C7H7ClO
142.582
MFCD00000557
QGRPVMLBTFGQDQ-UHFFFAOYSA-N
2-chloroanisole,o-chloroanisole,benzene, 1-chloro-2-methoxy,anisole, o-chloro,o-chloromethoxybenzene,o-chlorophenyl methyl ether,2-chlorophenol methyl ether,chloroanisole,1-chloro-2-methoxy-benzene,labotest-bb ltbb002921
13011
1-chloro-2-methoxybenzene
COC1=CC=CC=C1Cl
766-51-8
C7H7ClO
142.582
MFCD00000557
QGRPVMLBTFGQDQ-UHFFFAOYSA-N
2-chloroanisole,o-chloroanisole,benzene, 1-chloro-2-methoxy,anisole, o-chloro,o-chloromethoxybenzene,o-chlorophenyl methyl ether,2-chlorophenol methyl ether,chloroanisole,1-chloro-2-methoxy-benzene,labotest-bb ltbb002921
13011
1-chloro-2-methoxybenzene
COC1=CC=CC=C1Cl
Refractive Index 1.5435 to 1.5455 (20°C, 589nm)
Quantity 50g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H4OCH3
Beilstein 06, 184
Formula Weight 142.58
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloroanisole, 98%
Show More Show Less

EINECSNumber : 212-167-9

TSCA : TSCA