Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroanthracene 95%, Acros Organics

Click to view available options
Quantity:
250mg
17135-78-3
C14H9Cl
212.676
MFCD00003580
OWFINXQLBMJDJQ-UHFFFAOYSA-N
anthracene, 2-chloro,ccris 5549,.beta.-chloro anthracene,2-chloroanthracene,acmc-1c5s8,dsstox_cid_29086,dsstox_rid_83305,dsstox_gsid_49230,4-05-00-02292 beilstein handbook reference,owfinxqlbmjdjq-uhfffaoysa
28308
2-chloroanthracene
C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Cl
17135-78-3
C14H9Cl
212.676
MFCD00003580
OWFINXQLBMJDJQ-UHFFFAOYSA-N
anthracene, 2-chloro,ccris 5549,.beta.-chloro anthracene,2-chloroanthracene,acmc-1c5s8,dsstox_cid_29086,dsstox_rid_83305,dsstox_gsid_49230,4-05-00-02292 beilstein handbook reference,owfinxqlbmjdjq-uhfffaoysa
28308
2-chloroanthracene
C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Cl
Infrared Spectrum Authentic
Quantity 250mg
Color Beige to Beige-Brown
CAS Min % 94.0
CAS Max % 100.0
Formula Weight 212.68
Physical Form Powder or Flakes
Percent Purity ≥94%
Chemical Name or Material 2-Chloroanthracene, 95%