Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzhydrazide, 98+%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
5814-05-1
C7H7ClN2O
170.60
MFCD00007597
KPPNLSKVTKSSTG-UHFFFAOYSA-N
2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
79883
2-chlorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)Cl
5814-05-1
C7H7ClN2O
170.60
MFCD00007597
KPPNLSKVTKSSTG-UHFFFAOYSA-N
2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
79883
2-chlorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 116°C to 120°C
Beilstein 972229
Quantity 25g
Formula Weight 170.6
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzhydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 227-380-2

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only