Alfa Aesar™ 2-Chlorobenzimidazole, 97%

Manufacturer: Alfa Aesar™ A1219322

Quantity 100g

 View more versions of this product

Catalog No. AAA1219322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4857-06-1
    Molecular Formula C7H5ClN2
    Formula Weight 152.58
    MDL Number MFCD00051944
    Beilstein 116526
    Chemical Name or Material 2-Chlorobenzimidazole

    Specifications

    Formula Weight 152.58
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H5ClN2
    MDL Number MFCD00051944
    TSCA TSCA
    CAS 4857-06-1
    Beilstein 116526
    Health Hazard 1 Warning
    Melting Point ≈180° to 185°C
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Chlorobenzimidazole
    Assay 97%

    Description

    Applications
    2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit