Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzoic hydrazide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5814-05-1
C7H7ClN2O
170.60
MFCD00007597
KPPNLSKVTKSSTG-UHFFFAOYSA-N
2-chlorobenzhydrazide,2-chlorobenzoic hydrazide,2-chlorobenzoic acid hydrazide,2-chlorobenzene-1-carbohydrazide,benzoylhydrazine, o-chloro,benzoic acid, 2-chloro-, hydrazide,2-chlorobenzenecarbohydrazide,chlorobenzohydrazide,o-chlorobenzhydrazide,2clphcon2
79883
2-chlorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)Cl
5814-05-1
C7H7ClN2O
170.60
MFCD00007597
KPPNLSKVTKSSTG-UHFFFAOYSA-N
2-chlorobenzhydrazide,2-chlorobenzoic hydrazide,2-chlorobenzoic acid hydrazide,2-chlorobenzene-1-carbohydrazide,benzoylhydrazine, o-chloro,benzoic acid, 2-chloro-, hydrazide,2-chlorobenzenecarbohydrazide,chlorobenzohydrazide,o-chlorobenzhydrazide,2clphcon2
79883
2-chlorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)Cl
Infrared Spectrum Authentic
Quantity 25g
Color White to Beige
Melting Point 113°C to 118°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula ClC6H4CONHNH2
Formula Weight 170.6
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Chlorobenzoic hydrazide, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.