Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chlorobenzophenone, 99+%

Click to view available options
Quantity:
25g
100g
500g
5162-03-8
C13H9ClO
216.66
MFCD00000558
VMHYWKBKHMYRNF-UHFFFAOYSA-N
2-chlorobenzophenone,2-chlorophenyl phenyl methanone,methanone, 2-chlorophenyl phenyl,o-chlorobenzophenone,2-chlorophenyl phenylmethanone,benzophenone, 2-chloro,2-chlorophenyl-phenylmethanone,2-chlorobenyl phenyl ketone,2-chloro-phenyl-phenyl-methanone,benzophenone, 2-chloro-6ci,7ci,8ci
78838
(2-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl
5162-03-8
C13H9ClO
216.66
MFCD00000558
VMHYWKBKHMYRNF-UHFFFAOYSA-N
2-chlorobenzophenone,2-chlorophenyl phenyl methanone,methanone, 2-chlorophenyl phenyl,o-chlorobenzophenone,2-chlorophenyl phenylmethanone,benzophenone, 2-chloro,2-chlorophenyl-phenylmethanone,2-chlorobenyl phenyl ketone,2-chloro-phenyl-phenyl-methanone,benzophenone, 2-chloro-6ci,7ci,8ci
78838
(2-chlorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl
Boiling Point 330°C
Quantity 100g
Flash Point >110°C (230°F)
Melting Point 44°C to 47°C
Beilstein 1869594
Formula Weight 216.67
Percent Purity ≥99%
Chemical Name or Material 2-Chlorobenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 225-936-9

TSCA : Yes

Recommended Storage : Ambient temperatures