Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzothiazole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
615-20-3
C7H4ClNS
169.63
BSQLQMLFTHJVKS-UHFFFAOYSA-N
2-chlorobenzothiazole,benzothiazole, 2-chloro,2-chlorobenzo d thiazole,2-benzothiazolyl chloride,usaf ek-2784,2-chloro-benzothiazole,benzothiazole, chloro,unii-fn1t1y0p5p,fn1t1y0p5p,2-chlorobenzthiazole
11987
2-chloro-1,3-benzothiazole
C1=CC=C2C(=C1)N=C(S2)Cl
615-20-3
C7H4ClNS
169.63
BSQLQMLFTHJVKS-UHFFFAOYSA-N
2-chlorobenzothiazole,benzothiazole, 2-chloro,2-chlorobenzo d thiazole,2-benzothiazolyl chloride,usaf ek-2784,2-chloro-benzothiazole,benzothiazole, chloro,unii-fn1t1y0p5p,fn1t1y0p5p,2-chlorobenzthiazole
11987
2-chloro-1,3-benzothiazole
C1=CC=C2C(=C1)N=C(S2)Cl
Specific Gravity 1.37
Flash Point >110°C
Density 1.3700g/mL
Packaging Glass bottle
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Quantity 25g
Refractive Index 1.6367 to 1.6387
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 27, 44
Solubility Information Solubility in water: insoluble
Formula Weight 169.63
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzothiazole
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-415-