Alfa Aesar™ 2-Chlorobenzotrichloride, 99%

Manufacturer: Alfa Aesar™ B2442814

Quantity 25g

 View more versions of this product

Catalog No. AAB2442814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2136-89-2
    Molecular Formula C7H4Cl4
    Formula Weight 229.92
    MDL Number MFCD00000788
    UN Number UN3261
    Synonym alpha,alpha,alpha,2-Tetrachlorotoluene
    Chemical Name or Material 2-Chlorobenzotrichloride

    Specifications

    Formula Weight 229.92
    TSCA TSCA
    Molecular Formula C7H4Cl4
    CAS 2136-89-2
    UN Number UN3261
    Synonym alpha,alpha,alpha,2-Tetrachlorotoluene
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H351
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    Boiling Point 260° to 264°C
    Melting Point 28° to 31°C
    Quantity 25g
    MDL Number MFCD00000788
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2-Chlorobenzotrichloride
    Assay 99%

    GHS H Statement
    H301-H314-H318-H351
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit