Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chlorobenzotrifluoride, 99%

Manufacturer:  Alfa Aesar™ A1075336

Quantity 500g

 View more versions of this product

Catalog No. AAA1075336


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

88-16-4
180.56
UN2234
2-Chloro-alpha,alpha,alpha-trifluorotoluene
C7H4ClF3
MFCD00000561
510993
2-Chlorobenzotrifluoride

Specifications

58°C (136°F)
−6°C
88-16-4
180.56
UN2234
2-Chloro-alpha,alpha,alpha-trifluorotoluene
99%
151° to 153°C
500g
C7H4ClF3
MFCD00000561
510993
2-Chlorobenzotrifluoride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES

RTECSNumber : XS9141000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit