Learn More
2-Chlorobenzoxazole, 98%, Thermo Scientific Chemicals
Description
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Not miscible in water.
Notes
Store under inert gas. Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Keep away from heat.
Specifications
Density | 1.39 |
Boiling Point | 201°C to 202°C |
Flash Point | 85°C (185°F) |
Refractive Index | 1.567 |
Quantity | 5 g |
Beilstein | 115982 |
Solubility Information | Not miscible in water. |
Formula Weight | 153.57 |
Percent Purity | 99% |
Chemical Name or Material | 2-Chlorobenzoxazole |
Safety and Handling
Hazard Category | H227-H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H227 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
Precautionary Statement | P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P374-P501c |
EINECSNumber | 210-414-5 |
RTECSNumber | DM4680000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.