Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
1L
Packaging:
Glass bottle
609-65-4
C7H4Cl2O
175.008
ONIKNECPXCLUHT-UHFFFAOYSA-N
benzoyl chloride, 2-chloro,o-chlorobenzoyl chloride,chlorobenzoyl chloride,benzoyl chloride, o-chloro,ococ,2-chloro-benzoyl chloride,unii-jr29a4n74x,o-chlorobenzoylchloride,2-chlorobenzoylchloride,2-chlorobenzoic acid chloride
69110
CHEBI:60719
2-chlorobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)Cl
609-65-4
C7H4Cl2O
175.008
ONIKNECPXCLUHT-UHFFFAOYSA-N
benzoyl chloride, 2-chloro,o-chlorobenzoyl chloride,chlorobenzoyl chloride,benzoyl chloride, o-chloro,ococ,2-chloro-benzoyl chloride,unii-jr29a4n74x,o-chlorobenzoylchloride,2-chlorobenzoylchloride,2-chlorobenzoic acid chloride
69110
CHEBI:60719
2-chlorobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)Cl
Density 1.38g/mL
Quantity 5mL
Color Colorless to Yellow
Refractive Index 1.5708 to 1.5728 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula ClC6H4COCl
Formula Weight 175.01
Physical Form Liquid
Percent Purity ≥97.5% (GC)
Chemical Name or Material 2-Chlorobenzoyl chloride
Show More Show Less