Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzyl chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
611-19-8
C7H6Cl2
161.03
MFCD00000893
BASMANVIUSSIIM-UHFFFAOYSA-N
2-chlorobenzyl chloride,1-chloro-2-chloromethyl benzene,2-chlorobenzylchloride,o-chlorobenzyl chloride,alpha,2-dichlorotoluene,benzene, 1-chloro-2-chloromethyl,alpha,o-dichlorotoluene,ar,alpha-dichlorotoluene,ortho-alpha-dichlorotoluene,unii-h58di83rmd
11906
1-chloro-2-(chloromethyl)benzene
C1=CC=C(C(=C1)CCl)Cl
611-19-8
C7H6Cl2
161.03
MFCD00000893
BASMANVIUSSIIM-UHFFFAOYSA-N
2-chlorobenzyl chloride,1-chloro-2-chloromethyl benzene,2-chlorobenzylchloride,o-chlorobenzyl chloride,alpha,2-dichlorotoluene,benzene, 1-chloro-2-chloromethyl,alpha,o-dichlorotoluene,ar,alpha-dichlorotoluene,ortho-alpha-dichlorotoluene,unii-h58di83rmd
11906
1-chloro-2-(chloromethyl)benzene
C1=CC=C(C(=C1)CCl)Cl
Boiling Point 213°C to 214°C
Density 1.2700g/mL
Flash Point 82°C
Melting Point -13°C
Specific Gravity 1.27
Refractive Index 1.5580 to 1.56
Packaging Glass bottle
Color Colorless to Yellow
Quantity 250mL
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula ClC6H4CH2Cl
Beilstein 05, 297
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in most common solvents (reacts with man
Formula Weight 161.03
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 2-Chlorobenzyl chloride, 98+%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 210-258-8

RTECSNumber : CZ0195000

TSCA : TSCA