Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chlorobutyramide, 98%

Click to view available options
Quantity:
1g
5g
7462-73-9
C4H8ClNO
121.564
MFCD00041477
VJIOSCMTZVXQQU-UHFFFAOYSA-N
2-chlorobutyramide
344969
2-chlorobutanamide
CCC(C(=O)N)Cl
7462-73-9
C4H8ClNO
121.564
MFCD00041477
VJIOSCMTZVXQQU-UHFFFAOYSA-N
2-chlorobutyramide
344969
2-chlorobutanamide
CCC(C(=O)N)Cl
Melting Point 74°C to 76°C
Quantity 5g
Beilstein 1745706
Formula Weight 121.57
Percent Purity 98%
Chemical Name or Material 2-Chlorobutyramide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning