Alfa Aesar™ 2-Chlorocyclopentanone, 98%, stab. with potassium carbonate

Manufacturer: Alfa Aesar™ B2257306

Quantity 5g

 View more versions of this product

Catalog No. AAB2257306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 694-28-0
    Molecular Formula C5H7ClO
    Formula Weight 118.56
    MDL Number MFCD00001410
    Beilstein 1098899
    Chemical Name or Material 2-Chlorocyclopentanone, stabilized with potassium carbonate

    Specifications

    Formula Weight 118.56
    TSCA TSCA
    Molecular Formula C5H7ClO
    MDL Number MFCD00001410
    CAS 694-28-0
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    RTECS Number GY4900000
    Flash Point 77°C (171°F)
    Boiling Point 72° to 74°C (12mmHg)
    Quantity 5g
    Beilstein 1098899
    Chemical Name or Material 2-Chlorocyclopentanone, stabilized with potassium carbonate
    Assay 98%

    Description

    Applications
    2-Chlorocyclopentanone is used as pharmaceutical intermediate.

    Solubility
    Soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    −30° to −10°C

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit