Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloroethanesulfonyl chloride, 98%

Click to view available options
Quantity:
10g
50g
100g
250g
1622-32-8
C2H4Cl2O2S
163.012
MFCD00007461
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethyl sulphonyl chloride,2-chloroethylsulfonyl chloride,ethanesulfonyl chloride, 2-chloro,2-chloroethane sulfochloride,2-chloro-ethanesulfonyl chloride,2-chloroethanesulfonylchloride,beta-chloroethanesulfonyl chloride,2-chloroethane-1-sulfonyl chloride,ethanesulfonylchloride, 2-chloro,2-chloro-1-ethanesulfonyl chloride
15385
2-chloroethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl
1622-32-8
C2H4Cl2O2S
163.012
MFCD00007461
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethyl sulphonyl chloride,2-chloroethylsulfonyl chloride,ethanesulfonyl chloride, 2-chloro,2-chloroethane sulfochloride,2-chloro-ethanesulfonyl chloride,2-chloroethanesulfonylchloride,beta-chloroethanesulfonyl chloride,2-chloroethane-1-sulfonyl chloride,ethanesulfonylchloride, 2-chloro,2-chloro-1-ethanesulfonyl chloride
15385
2-chloroethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl
Applications
2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
Refractive Index 1.4915
Sensitivity Moisture sensitive
Density 1.56
Flash Point >110°C (230°F)
Quantity 10g
Boiling Point 203°C
Linear Formula ClCH2CH2SO2Cl
UN Number UN3390
Beilstein 1751202
Solubility Information Reacts with water.
Formula Weight 163.02
Percent Purity 98%
Chemical Name or Material 2-Chloroethanesulfonyl chloride
Show More Show Less

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H330-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 216-594-1

RTECSNumber : KI8060000

TSCA : Yes

Recommended Storage : Keep cold