Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethanol, 99+%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass bottle
107-07-3
C2H5ClO
80.51
MFCD00002829
SZIFAVKTNFCBPC-UHFFFAOYSA-N
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
34
CHEBI:28200
2-chloroethanol
C(CCl)O
107-07-3
C2H5ClO
80.51
MFCD00002829
SZIFAVKTNFCBPC-UHFFFAOYSA-N
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
34
CHEBI:28200
2-chloroethanol
C(CCl)O
Viscosity 3.4 mPa.s (20°C)
Acidity 0.2mg KOH/g max.
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point -63.0°C
Quantity 100mL
Flash Point 55°C
Specific Gravity 1.2
Density 1.2000g/mL
Water 0.1% max. (K.F.)
Color Colorless
Boiling Point 129.0°C
Refractive Index 1.4405 to 1.4425
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula ClCH2CH2OH
Beilstein 01, 337
Fieser 05,117
Merck Index 15, 3847
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,soluble in most organic liquids,insoluble in alifatic hydrocarbons
Formula Weight 80.51
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 2-Chloroethanol, 99+%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Fatal if swallowed.
Harmful in contact with skin.
Fatal if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE

GHS Signal Word: Danger

EINECSNumber : 203-459-7

RTECSNumber : KK0875000

TSCA : TSCA