Alfa Aesar™ 2-Chloroethyl ethyl ether, 98+%

Manufacturer: Alfa Aesar™ B2430814

Quantity 25g

 View more versions of this product

Catalog No. AAB2430814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 628-34-2
    Molecular Formula CH3CH2OCH2CH2Cl
    Formula Weight 108.57
    MDL Number MFCD00000974
    UN Number UN3271
    Synonym 1-Chloro-2-ethoxyethane; 2-Ethoxyethyl chloride
    Chemical Name or Material 2-Chloroethyl ethyl ether

    Specifications

    Formula Weight 108.57
    Molecular Formula CH3CH2OCH2CH2Cl
    MDL Number MFCD00000974
    CAS 628-34-2
    UN Number UN3271
    Synonym 1-Chloro-2-ethoxyethane; 2-Ethoxyethyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H331-H315-H319-H335
    Highly flammable liquid and vapour.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    Flash Point 6°C (43°F)
    Boiling Point 107° to 108°C
    Melting Point −60°C
    Quantity 25g
    Chemical Name or Material 2-Chloroethyl ethyl ether
    Assay ≥98%

    Description

    Applications
    It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.

    Solubility
    Miscible with water.

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H225-H331-H315-H319-H335
    Highly flammable liquid and vapour.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit