Alfa Aesar™ 2-Chloroethyl methanesulfonate, 99+%

Manufacturer: Alfa Aesar™ H5507903

Quantity 1g

 View more versions of this product

Catalog No. AAH5507903

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 3570-58-9
    Molecular Formula C3H7ClO3S
    Linear Formula CH3SO3CH2CH2Cl
    Formula Weight 158.6
    MDL Number MFCD00007560
    UN Number UN2810
    Synonym Methanesulfonic acid 2-chloroethyl ester
    Chemical Name or Material 2-Chloroethyl methanesulfonate

    Specifications

    Formula Weight 158.6
    MDL Number MFCD00007560
    Flash Point 113°C (235°F)
    Quantity 1g
    Molecular Formula C3H7ClO3S
    Linear Formula CH3SO3CH2CH2Cl
    UN Number UN2810
    Synonym Methanesulfonic acid 2-chloroethyl ester
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H351-H302-H312
    Toxic if inhaled.
    Suspected of causing cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    RTECS Number KK1960000
    Boiling Point 125° to 126°C (9mmHg)
    CAS 3570-58-9
    Chemical Name or Material 2-Chloroethyl methanesulfonate
    Assay ≥99%

    GHS H Statement
    H331-H351-H302-H312
    Toxic if inhaled.
    Suspected of causing cancer.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit