Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloroethyl methanesulfonate, 99+%

Manufacturer:  Alfa Aesar™ H5507914

Quantity 25g

 View more versions of this product

Catalog No. AAH5507914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

3570-58-9
CH3SO3CH2CH2Cl
MFCD00007560
Methanesulfonic acid 2-chloroethyl ester
C3H7ClO3S
158.6
UN2810
2-Chloroethyl methanesulfonate

Specifications

113°C (235°F)
25g
C3H7ClO3S
158.6
UN2810
2-Chloroethyl methanesulfonate
125° to 126°C (9mmHg)
3570-58-9
CH3SO3CH2CH2Cl
MFCD00007560
Methanesulfonic acid 2-chloroethyl ester
≥99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H351-H302-H312
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : KK1960000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit