Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl-p-toluenesulfonate 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
80-41-1
C9H11ClO3S
234.694
MFCD00000970
ZXNMIUJDTOMBPV-UHFFFAOYSA-N
2-chloroethyl p-toluenesulfonate,2-chloroethyl tosylate,2-chloroethyl p-tosylate,2-tosyloxyethyl chloride,2-chloroethanol p-toluenesulfonate,p-toluenesulfonic acid, 2-chloroethyl ester,ethanol, 2-chloro-, 4-methylbenzenesulfonate,2-p-toluenesulfonyloxy ethyl chloride,2-chloroethyl-p-toluenesulfonate,p-toluenesulfonic acid-2-chloroethyl ester
6639
2-chloroethyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCCCl
80-41-1
C9H11ClO3S
234.694
MFCD00000970
ZXNMIUJDTOMBPV-UHFFFAOYSA-N
2-chloroethyl p-toluenesulfonate,2-chloroethyl tosylate,2-chloroethyl p-tosylate,2-tosyloxyethyl chloride,2-chloroethanol p-toluenesulfonate,p-toluenesulfonic acid, 2-chloroethyl ester,ethanol, 2-chloro-, 4-methylbenzenesulfonate,2-p-toluenesulfonyloxy ethyl chloride,2-chloroethyl-p-toluenesulfonate,p-toluenesulfonic acid-2-chloroethyl ester
6639
2-chloroethyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCCCl
Packaging Glass bottle
Color Colorless to Yellow
Quantity 100g
Infrared Spectrum Authentic
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H4SO3CH2CH2Cl
Beilstein 11, II, 45
Formula Weight 234.7
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2-Chloroethyl-p-toluenesulfonate, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-277-2

RTECSNumber : XT6475000

TSCA : TSCA