Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloroethyl p-Toluenesulfonate 98.0+%, TCI America™
Click to view available options
Quantity:
25 g
100 g
Specifications
Color | Yellow |
Boiling Point | 156°C |
Quantity | 100 g |
Formula Weight | 234.69 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 2-Chloroethyl p-Toluenesulfonate |
Safety and Handling
RTECSNumber : XT6475000
TSCA : Yes