Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethoxy)ethyltrimethylsilane, tech. 90%, stab. with 0.1% N,N-Diisopropylethylamine, Thermo Scientific Chemicals

Catalog Number AAH5507403
Click to view available options
Quantity:
1 g
5 g
25 g
76513-69-4
C6H15ClOSi
166.72
MFCD00009919
BPXKZEMBEZGUAH-UHFFFAOYSA-N
2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
2724271
2-(chloromethoxy)ethyl-trimethylsilane
C[Si](C)(C)CCOCCl
76513-69-4
C6H15ClOSi
166.72
MFCD00009919
BPXKZEMBEZGUAH-UHFFFAOYSA-N
2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
2724271
2-(chloromethoxy)ethyl-trimethylsilane
C[Si](C)(C)CCOCCl

2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.

Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 170°C to 172°C
Quantity 1 g
Linear Formula (CH3)3SiCH2CH2OCH2Cl
UN Number UN2920
Beilstein 3587289
Flash Point 46°C (115°F)
Formula Weight 166.72
Percent Purity 90%
Grade Technical
Refractive Index 1.435
Sensitivity Moisture sensitive
Density 0.942
Chemical Name or Material 2-(Chloromethoxy)ethyltrimethylsilane, stabilized with 0.3% N-ethyldiisopropylamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 278-483-4
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit