Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Chloromethyl)benzothiazole, 95%

Click to view available options
Quantity:
250mg
1g
5g
37859-43-1
C8H6ClNS
183.653
MFCD05664944
SERUZNHRWBXDOX-UHFFFAOYSA-N
2-chloromethyl-1,3-benzothiazole,2-chloromethyl benzothiazole,2-chloromethyl benzo d thiazole,benzothiazole, 2-chloromethyl,2-chloromethylbenzothiazole,2-chloromethyl-benzothiazole,2-chloromethylbenzthiazole,acmc-1ag8v,ksc776s1l,2-chloromethylbenzo d thiazole
304978
2-(chloromethyl)-1,3-benzothiazole
C1=CC=C2C(=C1)N=C(S2)CCl
37859-43-1
C8H6ClNS
183.653
MFCD05664944
SERUZNHRWBXDOX-UHFFFAOYSA-N
2-chloromethyl-1,3-benzothiazole,2-chloromethyl benzothiazole,2-chloromethyl benzo d thiazole,benzothiazole, 2-chloromethyl,2-chloromethylbenzothiazole,2-chloromethyl-benzothiazole,2-chloromethylbenzthiazole,acmc-1ag8v,ksc776s1l,2-chloromethylbenzo d thiazole
304978
2-(chloromethyl)-1,3-benzothiazole
C1=CC=C2C(=C1)N=C(S2)CCl
Quantity 250mg
Melting Point 34°C
Sensitivity Moisture Sensitive
Boiling Point 88°C to 99°C (0.3mmHg)
Beilstein 120805
Formula Weight 183.65
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-(Chloromethyl)benzothiazole

storageNote1 : -30° to -10°C