Alfa Aesar™ 2-(Chloromethyl)pyridine hydrochloride, 98%

Manufacturer: Alfa Aesar™ A1022614

Quantity 25g

 View more versions of this product

Catalog No. AAA1022614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6959-47-3
    Molecular Formula C6H6ClN·HCl
    Formula Weight 164.04
    MDL Number MFCD00012811
    UN Number UN3261
    Beilstein 3689155
    Synonym 2-Picolyl chloride hydrochloride
    Chemical Name or Material 2- (Chloromethyl)pyridine hydrochloride

    Specifications

    Formula Weight 164.04
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 6959-47-3
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Molecular Formula C6H6ClN·HCl
    MDL Number MFCD00012811
    Health Hazard 1 Danger
    RTECS Number US6825000
    Beilstein 3689155
    Synonym 2-Picolyl chloride hydrochloride
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Melting Point 120° to 124°C
    Quantity 25g
    UN Number UN3261
    Chemical Name or Material 2- (Chloromethyl)pyridine hydrochloride
    Assay 98%

    Description

    Applications
    2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.

    Solubility
    Soluble in water, acetone (10-50mg/ml), 95%ethanol (100mg/ml).

    Notes
    Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H301-H311-H314-H318-H317
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit