Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloronicotinamide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
10366-35-5
C6H5ClN2O
156.57
MFCD00006237
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
2-chloronicotinamide,3-pyridinecarboxamide, 2-chloro,chloronicotinamide,2-chloro-nicotinamide,acmc-1bvtu,nicotinamide, 2-chloro,2-chloronicotinic acid amide,ksc492e7f,2-chloro-3-pyridinecarboxamide
82588
2-chloropyridine-3-carboxamide
C1=CC(=C(N=C1)Cl)C(=O)N
10366-35-5
C6H5ClN2O
156.57
MFCD00006237
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
2-chloronicotinamide,3-pyridinecarboxamide, 2-chloro,chloronicotinamide,2-chloro-nicotinamide,acmc-1bvtu,nicotinamide, 2-chloro,2-chloronicotinic acid amide,ksc492e7f,2-chloro-3-pyridinecarboxamide
82588
2-chloropyridine-3-carboxamide
C1=CC(=C(N=C1)Cl)C(=O)N
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Melting Point 162°C to 166°C
Color Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl(C5H3N)CONH2
Beilstein 22, II, 36
Formula Weight 156.57
Physical Form Fine Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Chloronicotinamide, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-808-9

TSCA : TSCA