Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenethyl bromide, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
16793-91-2
C8H8BrCl
219.51
AECBVDLERUETKG-UHFFFAOYSA-N
2-chlorophenethyl bromide,1-2-bromoethyl-2-chlorobenzene,2-2-chlorophenyl ethylbromide,1-2-bromo-ethyl-2-chloro-benzene,1-2-bromo-ethyl-2-chlorobenzene,benzene,1-2-bromoethyl-2-chloro,benzene, 1-2-bromoethyl-2-chloro,1-2-bromoethyl-2-chloro-benzene,2-chlorophenethylbromide,acmc-1cfee
2756963
1-(2-bromoethyl)-2-chlorobenzene
C1=CC=C(C(=C1)CCBr)Cl
16793-91-2
C8H8BrCl
219.51
AECBVDLERUETKG-UHFFFAOYSA-N
2-chlorophenethyl bromide,1-2-bromoethyl-2-chlorobenzene,2-2-chlorophenyl ethylbromide,1-2-bromo-ethyl-2-chloro-benzene,1-2-bromo-ethyl-2-chlorobenzene,benzene,1-2-bromoethyl-2-chloro,benzene, 1-2-bromoethyl-2-chloro,1-2-bromoethyl-2-chloro-benzene,2-chlorophenethylbromide,acmc-1cfee
2756963
1-(2-bromoethyl)-2-chlorobenzene
C1=CC=C(C(=C1)CCBr)Cl
Infrared Spectrum Authentic
Density 1.4569g/mL
Quantity 5g
Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 219.51
Physical Form Liquid
Percent Purity 95%
Specific Gravity 1.4569
Flash Point >110°C
Boiling Point 110°C to 112°C (10.0 Torr)
Chemical Name or Material 2-Chlorophenethyl bromide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Danger