Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenol, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
95-57-8
C6H5ClO
128.56
MFCD00002159
ISPYQTSUDJAMAB-UHFFFAOYSA-N
o-chlorophenol,phenol, 2-chloro,2-hydroxychlorobenzene,o-chlorphenol,o-chlorophenic acid,phenol, o-chloro,2-chloro-1-hydroxybenzene,chlorophenol,septi-kleen,2-chloro-phenol
7245
CHEBI:47083
2-chlorophenol
C1=CC=C(C(=C1)O)Cl
95-57-8
C6H5ClO
128.56
MFCD00002159
ISPYQTSUDJAMAB-UHFFFAOYSA-N
o-chlorophenol,phenol, 2-chloro,2-hydroxychlorobenzene,o-chlorphenol,o-chlorophenic acid,phenol, o-chloro,2-chloro-1-hydroxybenzene,chlorophenol,septi-kleen,2-chloro-phenol
7245
CHEBI:47083
2-chlorophenol
C1=CC=C(C(=C1)O)Cl
Quantity 10kg
Boiling Point 173.0°C to 175.0°C
Viscosity 4.11 mPa.s (25°C)
Melting Point 8.0°C
Packaging Plastic drum
Density 1.2620g/mL
Refractive Index 1.5580 to 1.5600
Flash Point 63°C
Specific Gravity 1.262
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula ClC6H4OH
Beilstein 06,183
Merck Index 15,2157
Solubility Information Solubility in water: 28.5g/l (20°C). Other solubilities: freely soluble in solutions of sodium hydroxide,, alcohol and ether, soluble in aromatic solvents
Formula Weight 128.56
Percent Purity 98+%
Chemical Name or Material 2-Chlorophenol, 98+%
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS Signal Word: Warning

EINECSNumber : 202-433-2

RTECSNumber : SK2625000

TSCA : TSCA