Alfa Aesar™ 2-Chlorophenol, 99%

Manufacturer: Alfa Aesar™ A1031130

Quantity 250g

 View more versions of this product

Catalog No. AAA1031130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95-57-8
    Molecular Formula C6H5ClO
    Formula Weight 128.56
    MDL Number MFCD00002159
    UN Number UN2021
    Beilstein 1905114
    Merck Index 14,2154
    Chemical Name or Material 2-Chlorophenol

    Specifications

    Formula Weight 128.56
    CAS 95-57-8
    Health Hazard 3 GHS P Statement
    P273-P302+P352
    Avoid release to the environment.
    IF ON SKIN: Wash with plenty of soap and water.
    RTECS Number SK2625000
    Flash Point 63°C (145°F)
    Melting Point 7° to 8°C
    TSCA TSCA
    UN Number UN2021
    Merck Index 14,2154
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.
    Odor Phenol-like
    Quantity 250g
    Molecular Formula C6H5ClO
    Beilstein 1905114
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, LIQUID
    Boiling Point 175° to 176°C
    MDL Number MFCD00002159
    Chemical Name or Material 2-Chlorophenol
    Assay 99%

    GHS H Statement
    H302-H312-H332-H227
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.

    GHS P Statement
    P273-P302+P352
    Avoid release to the environment.
    IF ON SKIN: Wash with plenty of soap and water.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit