Alfa Aesar™ 2-Chlorophenoxyacetonitrile, 97%

Manufacturer: Alfa Aesar™ A1134614

Quantity 25g

 View more versions of this product

Catalog No. AAA1134614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 43111-31-5
    Molecular Formula C8H6ClNO
    Formula Weight 167.6
    MDL Number MFCD00017331
    UN Number UN3276
    Beilstein 2089865
    Chemical Name or Material 2-Chlorophenoxyacetonitrile

    Specifications

    Formula Weight 167.6
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00017331
    CAS 43111-31-5
    Quantity 25g
    UN Number UN3276
    Beilstein 2089865
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Molecular Formula C8H6ClNO
    Health Hazard 1 Danger
    Boiling Point 109° to 110°C (1mmHg)
    Chemical Name or Material 2-Chlorophenoxyacetonitrile
    Assay 97%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit