Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenylacetic acid, 98.5+%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2444-36-2
C8H7ClO2
170.6
MFCD00004317
IUJAAIZKRJJZGQ-UHFFFAOYSA-N
2-chlorophenylacetic acid,o-chlorophenylacetic acid,2-2-chlorophenyl acetic acid,2-chlorophenyl acetic acid,2-chlorophenylaceticacid,benzeneacetic acid, 2-chloro,o-chlorophenyl acetic acid,acetic acid, o-chlorophenyl,2-chloro-benzeneacetic acid
17124
2-(2-chlorophenyl)acetic acid
C1=CC=C(C(=C1)CC(=O)O)Cl
2444-36-2
C8H7ClO2
170.6
MFCD00004317
IUJAAIZKRJJZGQ-UHFFFAOYSA-N
2-chlorophenylacetic acid,o-chlorophenylacetic acid,2-2-chlorophenyl acetic acid,2-chlorophenyl acetic acid,2-chlorophenylaceticacid,benzeneacetic acid, 2-chloro,o-chlorophenyl acetic acid,acetic acid, o-chlorophenyl,2-chloro-benzeneacetic acid
17124
2-(2-chlorophenyl)acetic acid
C1=CC=C(C(=C1)CC(=O)O)Cl
Packaging Glass bottle
Melting Point 94.0°C to 97.0°C
Quantity 100g
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5+%
Linear Formula ClC6H4CH2CO2H
Beilstein 09, 447
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 170.6
Physical Form Powder
Percent Purity 98.5+%
Chemical Name or Material 2-Chlorophenylacetic acid
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 219-482-