Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chlorophenylboronic acid, 97%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Color | White |
Quantity | 1g |
Infrared Spectrum | Authentic |
Melting Point | 94.0°C to 95.0°C |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Assay Percent Range | 96% min. (HPLC) |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol |
Formula Weight | 156.38 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning