Alfa Aesar™ 2-Chloropyrazine, 98%

Manufacturer: Alfa Aesar™ A1010814

Quantity 25g

 View more versions of this product

Catalog No. AAA1010814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14508-49-7
    Molecular Formula C4H3ClN2
    Formula Weight 114.54
    MDL Number MFCD00006124
    UN Number UN1993
    Beilstein 107019
    Chemical Name or Material 2-Chloropyrazine

    Specifications

    Formula Weight 114.54
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H3ClN2
    UN Number UN1993
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Boiling Point 153° to 154°C
    TSCA TSCA
    CAS 14508-49-7
    Beilstein 107019
    Flash Point 57°C (134°F)
    MDL Number MFCD00006124
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 25g
    Chemical Name or Material 2-Chloropyrazine
    Assay 98%

    Description

    Applications
    2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

    Solubility
    Soluble in Methanol

    Notes
    Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit