Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloropyridine-3-sulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Acros Organics 434310010

 View more versions of this product

Catalog No. AC434310010

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

2-Chloropyridine-3-sulfonyl chloride
Glass bottle
42.0°C to 53.0°C
6686-02-8
100.0
C20H21N3O3
IMMPMHKLUUZKAZ-DAFXYXGESA-N
(2S)-2-[[(2S)-2-amino-3-(1H-indol-3-yl)propanoyl]amino]-3-phenylpropanoic acid
6426942
212.06
≥96.0% diethylsulfonamide derivative (HPLC)
Authentic
White
1g
96.0
97%
2s-2-2s-2-amino-3-1h-indol-3-yl propanamido-3-phenylpropanoic acid, 2s-2-2s-2-amino-3-1h-indol-3-yl propanoyl amino-3-phenylpropanoic acid, h-trp-phe-oh, l-phenylalanine, l-tryptophyl, l-trp-l-phe, l-trp-l-phe-oh, l-tryptophyl-l-phenylalanine, s-2-s-2-amino-3-1h-indol-3-yl propanamido-3-phenylpropanoic acid, tryptophyl-phenylalanine, wf
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)C(CC2=CNC3=CC=CC=C32)N
351.41
CHEBI:74874
Powder
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTRE or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit