Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropyridine-4-boronic acid pinacol ester, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
458532-84-8
C11H15BClNO2
239.51
UUEQDBHKMOFLDP-UHFFFAOYSA-N
2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-chloropyridine-4-boronic acid pinacol ester,2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-chloropyridin-4-ylboronic acid pinacol ester,2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine,2-chloropyridine-4-boronic acid, pinacol ester,2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine,2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine,pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,pubchem16684
5103408
2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2)Cl
458532-84-8
C11H15BClNO2
239.51
UUEQDBHKMOFLDP-UHFFFAOYSA-N
2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-chloropyridine-4-boronic acid pinacol ester,2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine,2-chloropyridin-4-ylboronic acid pinacol ester,2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine,2-chloropyridine-4-boronic acid, pinacol ester,2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine,2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine,pyridine, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,pubchem16684
5103408
2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2)Cl
Packaging Glass Bottle
Quantity 1g
Melting Point 63.5°C to 66.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Formula Weight 239.51
Percent Purity 97%
Chemical Name or Material 2-Chloropyridine-4-boronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Warning